missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7128363
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46031G 1 g
D46035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46031G Supplier TCI America Supplier No. D46031G
Only null left

Chemical Identifiers

CAS 2510-36-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00051657
InChI Key IJEUISLJVBUNRE-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
PubChem CID 75636
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C)C(=O)O

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 1 g
Formula Weight 141.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethylisoxazole-4-carboxylic Acid