missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carboxylic acid, 99%

Catalog No. AAA1541906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1541906 5 g
AAA1541914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1541906 Supplier Thermo Scientific Chemicals Supplier No. A1541906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2510-36-3 | C6H7NO3 | 141.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2510-36-3
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00051657
InChI Key IJEUISLJVBUNRE-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carboxylic acid, 3,5-dimethylisoxasole-4-carboxylic acid, 4-isoxazolecarboxylic acid, 3,5-dimethyl, dimethyl-1,2-oxazole-4-carboxylic acid, 3,5-dimethyl-4-isoxazolecarboxylic acid, 3,5-dimethyl-isoxazole-4-carboxylic acid, 3,5-dimethylisoxazole-4-carboxylicacid, pubchem8774, akos pao-1555
PubChem CID 75636
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(=NO1)C)C(=O)O

Specifications

Melting Point 140°C to 142°C
Quantity 5 g
Beilstein 115819
Formula Weight 141.13
Percent Purity 99%
Chemical Name or Material 3,5-Dimethylisoxazole-4-carboxylic acid
EINECSNumber 219-724-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only