missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-carbonyl Chloride 96.0+%, TCI America™

Catalog No. D47061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47061G 1 g
D47065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47061G Supplier TCI America Supplier No. D47061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 31301-45-8
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.569
MDL Number MFCD00052555
InChI Key MPYGFFPGJMGVSW-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-carbonyl chloride, 4-isoxazolecarbonyl chloride, 3,5-dimethyl, 3,5-dimethylisoxazole-4-carbonylchloride, dimethyl-1,2-oxazole-4-carbonyl chloride, 3,5-dimethyl-4-isoxazolecarbonyl chloride, pubchem8669, acmc-20a3wc, 3,5-dimethylisoxasole-4-carbonyl chloride, 3,5-dimethyisoxazole-4-carbonyl chloride, 3,5 dimethylisoxazole-4-carbonyl chloride
PubChem CID 2736265
IUPAC Name 3,5-dimethyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C(=NO1)C)C(=O)Cl

Specifications

Color Yellow
Boiling Point 90°C
Quantity 1 g
UN Number 3265
Formula Weight 159.57
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylisoxazole-4-carbonyl Chloride