missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-boronic acid pinacol ester, 97%

Catalog No. p-7056874
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6604206 5 g
AAH6604214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6604206 Supplier Thermo Scientific Chemicals Supplier No. H6604206
Only null left

CAS: 832114-00-8 | C11H18BNO3 | 223.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 832114-00-8
Molecular Formula C11H18BNO3
Molecular Weight (g/mol) 223.079
MDL Number MFCD05863910
InChI Key CVLHETBAROWASE-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid pinacol ester, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid, pinacol ester, 3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-isoxazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3,5-dimethylisoxazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pubchem18439, acmc-209prk
PubChem CID 2758656
IUPAC Name 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(ON=C2C)C

Specifications

Melting Point 95°C to 99°C
Color Brown to White
Quantity 5 g
Formula Weight 223.08
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3,5-Dimethylisoxazole-4-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only