Learn More
3,5-Dimethylisoxazole-4-boronic acid, 97%
CAS: 16114-47-9 | C5H8BNO3 | 140.933 g/mol
$192.42 - $571.96
Chemical Identifiers
| CAS | 16114-47-9 |
|---|---|
| Molecular Formula | C5H8BNO3 |
| Molecular Weight (g/mol) | 140.933 |
| MDL Number | MFCD02677945 |
| InChI Key | DIIFZCPZIRQDIJ-UHFFFAOYSA-N |
| Synonym | 3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid |
| PubChem CID | 2734346 |
| IUPAC Name | (3,5-dimethyl-1,2-oxazol-4-yl)boronic acid |
| SMILES | B(C1=C(ON=C1C)C)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2689303
|
Thermo Scientific Chemicals
H2689303 |
1 g |
Each for $192.42
|
|
|||||
|
AAH2689306
|
Thermo Scientific Chemicals
H2689306 |
5 g |
Each for $571.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 16114-47-9 | |
| 140.933 | |
| DIIFZCPZIRQDIJ-UHFFFAOYSA-N | |
| 2734346 | |
| B(C1=C(ON=C1C)C)(O)O |
| C5H8BNO3 | |
| MFCD02677945 | |
| 3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid | |
| (3,5-dimethyl-1,2-oxazol-4-yl)boronic acid |
Specifications
| 16114-47-9 | |
| C5H8BNO3 | |
| 1 g | |
| DIIFZCPZIRQDIJ-UHFFFAOYSA-N | |
| (3,5-dimethyl-1,2-oxazol-4-yl)boronic acid | |
| 2734346 | |
| 97% |
| 106°C to 108°C | |
| MFCD02677945 | |
| 3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid | |
| B(C1=C(ON=C1C)C)(O)O | |
| 140.933 | |
| 140.93 | |
| 3,5-Dimethylisoxazole-4-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only