missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazole-4-boronic acid, 97%

Catalog No. AAH2689303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2689303 1 g
AAH2689306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2689303 Supplier Thermo Scientific Chemicals Supplier No. H2689303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16114-47-9 | C5H8BNO3 | 140.933 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16114-47-9
Molecular Formula C5H8BNO3
Molecular Weight (g/mol) 140.933
MDL Number MFCD02677945
InChI Key DIIFZCPZIRQDIJ-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazole-4-boronic acid, 3,5-dimethylisoxazol-4-yl boronic acid, 3,5-dimethyl-4-isoxazolylboronic acid, 3,5-dimethylisoxazol-4-ylboronic acid, 3,5-dimethyl-1,2-oxazol-4-yl boronic acid, 3,5-dimethylisoxazole-4-boronicacid, 3,5-dimethyl-4-isoxazoleboronic acid, 3,5-dimethylisoxazol-4-yl-4-boronic acid, 3,5-dimethyl-isoxazole-4-boronic acid, dimethyl-1,2-oxazol-4-yl boronic acid
PubChem CID 2734346
IUPAC Name (3,5-dimethyl-1,2-oxazol-4-yl)boronic acid
SMILES B(C1=C(ON=C1C)C)(O)O

Specifications

Melting Point 106°C to 108°C
Quantity 1 g
Formula Weight 140.93
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylisoxazole-4-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only