missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylisoxazol-4-yl isocyanate, 95%, Thermo Scientific™

Catalog No. CC00606CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00606CB 250 mg Amber Glass Bottle
CC00606DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC00606CB Supplier Thermo Scientific Chemicals Supplier No. CC00606CB

Chemical Identifiers

CAS 131825-41-7
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00119910
InChI Key JJQYAGYVYNMYGE-UHFFFAOYSA-N
Synonym 3,5-dimethylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3,5-dimethyl, 3,5-dimethylisoxazole-4-isocyanate, 4-isocyanato-3,5-dimethylisoxazole, 3,5-dimethyl isoxazol-4-yl isocyanate, 3,5-dimethylisoxazol-4-ylisocyanate, acmc-20mu97, 3,5-dimethylisoxazol-4-isocyanate, 3,5-dimethyl-4-isocyanatoisoxazole, 4-isocyanato3,5-dimethyl-isoxazole
PubChem CID 2736266
IUPAC Name 4-isocyanato-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)N=C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 138.13
Percent Purity 95%
Chemical Name or Material 3, 5-Dimethylisoxazol-4-yl isocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.