missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylcyclohexanone (mixture of isomers) 98.0+%, TCI America™

$82.02 - $415.68

Chemical Identifiers

CAS 2320-30-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00019467
InChI Key MSANHHHQJYQEOK-UHFFFAOYSA-N
PubChem CID 137544
IUPAC Name 3,5-dimethylcyclohexan-1-one
SMILES CC1CC(CC(=O)C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D12825ML
View Documents
TCI America
D12825ML
5 mL N/A
Only null left
 
D128225ML
View Documents
TCI America
D128225ML
25 mL N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2320-30-1
126.199
MSANHHHQJYQEOK-UHFFFAOYSA-N
3,5-dimethylcyclohexan-1-one
C8H14O
MFCD00019467
137544
CC1CC(CC(=O)C1)C

Specifications

2320-30-1
176°C
MFCD00019467
1224
CC1CC(CC(=O)C1)C
126.199
126.20
Liquid
Yellow
C8H14O
5 mL
MSANHHHQJYQEOK-UHFFFAOYSA-N
3,5-dimethylcyclohexan-1-one
137544
≥98.0% (GC)
3,5-Dimethylcyclohexanone (mixture of isomers)
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents