missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylbenzo[b]thiophene, 97%

Catalog No. p-7025339
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2076803 1 g
AAB2076806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2076803 Supplier Thermo Scientific Chemicals Supplier No. B2076803

CAS: 1964-45-0 | C10H10S | 162.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1964-45-0
Molecular Formula C10H10S
Molecular Weight (g/mol) 162.25
MDL Number MFCD00052744
InChI Key UQJSHXYNNVEGMQ-UHFFFAOYSA-N
Synonym 3,5-dimethylbenzo b thiophene, 3,5-dimethyl-benzo b thiophene, 3,5-dimethylbenzothiophene, 3,5-dimethylthianapthene, benzo b thiophene, 3,5-dimethyl, 3,5-dimethyl benzo b thiophene, 3,5-dimethyl benzothiophene
PubChem CID 2779875
IUPAC Name 3,5-dimethyl-1-benzothiophene
SMILES CC1=CC2=C(C=C1)SC=C2C

Specifications

Boiling Point 62°C to 64°C (0.1 mmHg)
Quantity 1 g
Formula Weight 162.25
Percent Purity 97%
Chemical Name or Material 3,5-Dimethylbenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only