missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyladamantane-1-carboxylic acid, 97%

Catalog No. AAH2707303
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2707303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2707303 Supplier Thermo Scientific Chemicals Supplier No. H2707303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14670-94-1 | C13H19O2 | 207.29 g/mol

3,5-Dimethyladamantane-1-carboxylic acid, is used as a pharmaceutical and API intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyladamantane-1-carboxylic acid, is used as a pharmaceutical and API intermediate.

Solubility
Sparingly Soluble (0.14 g/L) (25°C)

Notes
Store in cool place. Keep container tightly close

Chemical Identifiers

CAS 14670-94-1
Molecular Formula C13H19O2
Molecular Weight (g/mol) 207.29
MDL Number MFCD00188067
InChI Key BSWOQWGHXZTDOO-QGVZIFMBSA-M
Synonym 3,5-dimethyl-1-adamantanecarboxylic acid, 1,3-dimethyladamantan-5-carboxylic acid, 3,5-dimethyladamantanecarboxylic acid, maybridge1_002240, 3,5-dimethyltricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3,5-dimethyl adamantane-1-carboxylic acid, acmc-20ancb, 3,5-dimethyladamantane carboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3,5-dimethyl
PubChem CID 2736224
IUPAC Name 3,5-dimethyladamantane-1-carboxylic acid
SMILES C[C@@]12CC3C[C@@](C)(C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 107°C to 109°C
Quantity 1 g
Solubility Information Sparingly Soluble (0.14g/L) (25°C)
Formula Weight 208.3
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyladamantane-1-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only