missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole 98.0+%, TCI America™

Catalog No. D37721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37721G 1 g
D37725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37721G Supplier TCI America Supplier No. D37721G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 832114-00-8
Molecular Formula C11H18BNO3
Molecular Weight (g/mol) 223.079
MDL Number MFCD05863910
InChI Key CVLHETBAROWASE-UHFFFAOYSA-N
Synonym 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid pinacol ester, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl isoxazole, 3,5-dimethylisoxazole-4-boronic acid, pinacol ester, 3,5-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,2-oxazole, 3,5-dimethyl-4-isoxazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-3,5-dimethylisoxazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pubchem18439, acmc-209prk
PubChem CID 2758656
IUPAC Name 3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(ON=C2C)C

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole