missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-2-pyrrolecarboxaldehyde 98.0+%, TCI America™
$212.51 - $726.98
Chemical Identifiers
| CAS | 2199-58-8 |
|---|---|
| Molecular Formula | C7H9NO |
| Molecular Weight (g/mol) | 123.155 |
| MDL Number | MFCD00111522 |
| InChI Key | RDFZYUOHJBXMJA-UHFFFAOYSA-N |
| Synonym | 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole |
| PubChem CID | 270465 |
| IUPAC Name | 3,5-dimethyl-1H-pyrrole-2-carbaldehyde |
| SMILES | CC1=CC(=C(N1)C=O)C |
Chemical Identifiers
| 2199-58-8 | |
| 123.155 | |
| RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
| 270465 | |
| CC1=CC(=C(N1)C=O)C |
| C7H9NO | |
| MFCD00111522 | |
| 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
| 3,5-dimethyl-1H-pyrrole-2-carbaldehyde |
Specifications
| 2199-58-8 | |
| C7H9NO | |
| 1 g | |
| RDFZYUOHJBXMJA-UHFFFAOYSA-N | |
| 3,5-dimethyl-1H-pyrrole-2-carbaldehyde | |
| 270465 | |
| ≥98.0% (GC) | |
| 3,5-Dimethyl-2-pyrrolecarboxaldehyde |
| 91°C | |
| MFCD00111522 | |
| 3,5-dimethylpyrrole-2-carboxaldehyde, 3,5-dimethyl-1h-pyrrole-2-carboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 3,5-dimethyl, 3,5-dimethyl-2-pyrrolecarboxaldehyde, pubchem2127, acmc-209fqe, ksc202o9r, 3,5-dimethyl-2-formylpyrrol, 2,4-dimethyl-5-formylpyrrole, 2-formyl-3,5-dimethylpyrrole | |
| CC1=CC(=C(N1)C=O)C | |
| 123.155 | |
| 123.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No