missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-1-hexyn-3-ol 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 150°C |
| Quantity | 25 mL |
| UN Number | 1987 |
| Formula Weight | 126.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3,5-Dimethyl-1-hexyn-3-ol |
Safety and Handling
| EINECSNumber | (2)-0265 |
| TSCA | Yes |