missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-hexyn-3-ol 98.0+%, TCI America™

Catalog No. D073725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D073725ML 25 mL
D0737500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D073725ML Supplier TCI America Supplier No. D073725ML
Only null left

Chemical Identifiers

CAS 107-54-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00008941
InChI Key NECRQCBKTGZNMH-UHFFFAOYNA-N
Synonym 3,5-dimethyl-1-hexyn-3-ol, surfynol, surfynol 61, 1-hexyn-3-ol, 3,5-dimethyl, surfynol van, sialyl-3-paragloboside, acmc-2098wy, dsstox_cid_21472, dsstox_rid_79746, dsstox_gsid_41472
PubChem CID 61018
IUPAC Name 3,5-dimethylhex-1-yn-3-ol
SMILES CC(C)CC(C)(O)C#C

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
UN Number 1987
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethyl-1-hexyn-3-ol
EINECSNumber (2)-0265
TSCA Yes