missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1-adamantanol 97.0+%, TCI America™

Catalog No. p-7127870
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36971G 1 g
D36975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36971G Supplier TCI America Supplier No. D36971G

Chemical Identifiers

CAS 707-37-9
Molecular Formula C12H20O
Molecular Weight (g/mol) 180.291
MDL Number MFCD00074775
InChI Key LBWCITVBZLTEKW-UHFFFAOYSA-N
Synonym 1-Hydroxy-3,5-dimethyladamantane
PubChem CID 265793
IUPAC Name 3,5-dimethyladamantan-1-ol
SMILES CC12CC3CC(C1)(CC(C3)(C2)O)C

Specifications

Melting Point 95°C
Color White
Quantity 1 g
Formula Weight 180.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-1-adamantanol