missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethyl-1-adamantanamine Hydrochloride 98.0+%, TCI America™
$69.31 - $273.38
Chemical Identifiers
| CAS | 41100-52-1 |
|---|---|
| Molecular Formula | C12H22ClN |
| Molecular Weight (g/mol) | 215.77 |
| MDL Number | MFCD00214336 |
| InChI Key | LDDHMLJTFXJGPI-UHFFFAOYNA-N |
| Synonym | memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl |
| PubChem CID | 181458 |
| ChEBI | CHEBI:64323 |
| IUPAC Name | hydrogen 3,5-dimethyladamantan-1-amine chloride |
| SMILES | [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 |
Chemical Identifiers
| 41100-52-1 | |
| 215.77 | |
| LDDHMLJTFXJGPI-UHFFFAOYNA-N | |
| 181458 | |
| hydrogen 3,5-dimethyladamantan-1-amine chloride |
| C12H22ClN | |
| MFCD00214336 | |
| memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl | |
| CHEBI:64323 | |
| [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 |
Specifications
| 41100-52-1 | |
| White | |
| MFCD00214336 | |
| memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl | |
| [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 | |
| 215.77 | |
| CHEBI:64323 | |
| ≥98.0% (N,T) | |
| 3,5-Dimethyl-1-adamantanamine Hydrochloride |
| 292°C | |
| C12H22ClN | |
| 5 g | |
| LDDHMLJTFXJGPI-UHFFFAOYNA-N | |
| hydrogen 3,5-dimethyladamantan-1-amine chloride | |
| 181458 | |
| 215.77 | |
| Crystalline Powder |
Safety and Handling
TSCA : No