missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxybenzaldehyde, 98%

Catalog No. p-7022367
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1510906 5 g
AAA1510914 25 g
AAA1510922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1510906 Supplier Thermo Scientific Chemicals Supplier No. A1510906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7311-34-4 | C9H10O3 | 166.18 g/mol

3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.

Solubility
Soluble in hot methanol. Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.

Chemical Identifiers

CAS 7311-34-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003366
InChI Key VFZRZRDOXPRTSC-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-benzaldehyde, benzaldehyde, 3,5-dimethoxy, 3,5-dimethoxybenzaldehyd, pubchem2645, 3,5 dimethoxybenzaldehyde, acmc-1bhx6, 3,5 dimethoxylbenzaldehyde, 3,5 dimethoxy benzaldehyde, 3,5-dimethoxy benzaldehyde, ghl.pd_mitscher_leg0.694
PubChem CID 81747
IUPAC Name 3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1

Specifications

Melting Point 44°C to 48°C
Boiling Point 115°C to 118°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2042374
Sensitivity Air Sensitive
Solubility Information Soluble in hot methanol. Insoluble in water.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 3,5-Dimethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-772-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only