missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethoxybenzaldehyde, 98%
Description
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.
Solubility
Soluble in hot methanol. Insoluble in water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
3,5-Dimethoxybenzaldehyde is used as a building block in organic synthesis. It is used to prepare isomeric hyperbranched polyetherketones.
Solubility
Soluble in hot methanol. Insoluble in water.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
Specifications
| Melting Point | 44°C to 48°C |
| Boiling Point | 115°C to 118°C (1 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 25 g |
| Beilstein | 2042374 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in hot methanol. Insoluble in water. |
| Formula Weight | 166.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 3,5-Dimethoxybenzaldehyde |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 230-772-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only