missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-hydroxycinnamic Acid 98.0+%, TCI America™

Catalog No. p-7126917
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17655G 5 g
D176525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17655G Supplier TCI America Supplier No. D17655G

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD00004401
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Specifications

Melting Point 198°C
Quantity 5 g
Formula Weight 224.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxy-4-hydroxycinnamic Acid