missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethoxy-4-hydroxycinnamic acid, 98%, predominantly trans isomer

Catalog No. p-4258967
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115550010 1 g Glass bottle
AC115550050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115550010 Supplier Thermo Scientific Chemicals Supplier No. 115550010

CAS: 530-59-6 | C11H12O5 | 224.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.21
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 203.0°C to 205.0°C
Color Beige-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H2(OCH3)2CH=CHCO2H
Quantity 1 g
Beilstein 10,508
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in methanol,ethanol and acetone
Formula Weight 224.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dimethoxy-4-hydroxycinnamic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-487-3

RUO – Research Use Only