missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylaldehyde 98.0+%, TCI America™

Catalog No. D48485G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48485G Supplier TCI America Supplier No. D48485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.916
MDL Number MFCD00003321
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I

Specifications

Melting Point 109°C
Quantity 5 g
Formula Weight 373.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodosalicylaldehyde