Learn More
3,5-Diiodosalicylaldehyde, 98+%
CAS: 2631-77-8 | C7H4I2O2 | 373.916 g/mol
Supplier: Thermo Scientific Chemicals L0578318
Description
3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N?-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N′-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.
Solubility
Insoluble in water.
Notes
Air & Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.
Specifications
| 3,5-Diiodosalicylaldehyde | |
| 106°C to 110°C | |
| MFCD00003321 | |
| 1102552 | |
| 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo | |
| MYWSBJKVOUZCIA-UHFFFAOYSA-N | |
| 2-hydroxy-3,5-diiodobenzaldehyde | |
| 75829 | |
| ≥98% |
| 2631-77-8 | |
| C7H4I2O2 | |
| 50 g | |
| Air and light sensitive | |
| Insoluble in water. | |
| C1=C(C=C(C(=C1I)O)C=O)I | |
| 373.916 | |
| 373.92 |
Chemical Identifiers
| 2631-77-8 | |
| 373.916 | |
| MYWSBJKVOUZCIA-UHFFFAOYSA-N | |
| 2-hydroxy-3,5-diiodobenzaldehyde |
| C7H4I2O2 | |
| MFCD00003321 | |
| 75829 | |
| C1=C(C=C(C(=C1I)O)C=O)I |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-117-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only