missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylaldehyde, 98+%

Catalog No. AAL0578309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0578309 10 g
AAL0578318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0578309 Supplier Thermo Scientific Chemicals Supplier No. L0578309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2631-77-8 | C7H4I2O2 | 373.916 g/mol

3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N?-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diiodosalicylaldehyde is used to synthesize new Schiff bases, (2,4-diiodo-6-[(2-morpholin-4-yl-ethylimino)-methyl]-phenol and 2,4-diiodo-6-[(3-morpholin-4-yl-propylimino)-methyl]-phenol) and the new tridentate ligand, [(2-hydroxy-3,5-diiodo-benzylidene)-amino]-acetic acid (HDBA), 3-bromo-N′-(2-hydroxy-3,5-diiodobenzylidene)benzohydrazide monohydrate.

Solubility
Insoluble in water.

Notes
Air & Light Sensitive. Store in the dark. Incompatible with light and oxidizing agents.

Chemical Identifiers

CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.916
MDL Number MFCD00003321
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I

Specifications

Melting Point 106°C to 110°C
Quantity 10 g
Beilstein 1102552
Sensitivity Air and light sensitive
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
Solubility Information Insoluble in water.
Formula Weight 373.92
Percent Purity ≥98%
Chemical Name or Material 3,5-Diiodosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-117-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only