missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diiodosalicylaldehyde, 97%
CAS: 2631-77-8 | C7H4I2O2 | 373.92 g/mol
$330.40 - $330.40
Chemical Identifiers
| CAS | 2631-77-8 |
|---|---|
| Molecular Formula | C7H4I2O2 |
| Molecular Weight (g/mol) | 373.92 |
| InChI Key | MYWSBJKVOUZCIA-UHFFFAOYSA-N |
| Synonym | 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo |
| PubChem CID | 75829 |
| IUPAC Name | 2-hydroxy-3,5-diiodobenzaldehyde |
| SMILES | C1=C(C=C(C(=C1I)O)C=O)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC450860250
|
Thermo Scientific Chemicals
450860250 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2631-77-8 | |
| 373.92 | |
| 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo | |
| 2-hydroxy-3,5-diiodobenzaldehyde |
| C7H4I2O2 | |
| MYWSBJKVOUZCIA-UHFFFAOYSA-N | |
| 75829 | |
| C1=C(C=C(C(=C1I)O)C=O)I |
Specifications
| 2631-77-8 | |
| 96% min. (GC) | |
| 25 g | |
| MYWSBJKVOUZCIA-UHFFFAOYSA-N | |
| 2-hydroxy-3,5-diiodobenzaldehyde | |
| 75829 | |
| 97% |
| 108°C to 110°C | |
| C7H4I2O2 | |
| 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo | |
| C1=C(C=C(C(=C1I)O)C=O)I | |
| 373.92 | |
| 373.92 | |
| 3, 5-Diiodosalicylaldehyde |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 220-117-2
RUO – Research Use Only