missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodosalicylaldehyde, 97%

Catalog No. AC450860050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450860050 5 g
AC450860250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450860050 Supplier Thermo Scientific Chemicals Supplier No. 450860050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2631-77-8 | C7H4I2O2 | 373.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2631-77-8
Molecular Formula C7H4I2O2
Molecular Weight (g/mol) 373.92
InChI Key MYWSBJKVOUZCIA-UHFFFAOYSA-N
Synonym 3,5-diiodosalicylaldehyde, benzaldehyde, 2-hydroxy-3,5-diiodo, 3,5-diiodo-2-hydroxybenzaldehyde, 2-hydroxy-3,5-diiodo-benzaldehyde, acmc-20ancp, 3,5-diodosolicylaldehyde, 2,4-diiodo-6-formylphenol, #, benzaldehyde,2-hydroxy-3,5-diiodo
PubChem CID 75829
IUPAC Name 2-hydroxy-3,5-diiodobenzaldehyde
SMILES C1=C(C=C(C(=C1I)O)C=O)I

Specifications

Melting Point 108°C to 110°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 373.92
Percent Purity 97%
Chemical Name or Material 3, 5-Diiodosalicylaldehyde
Hazard Category GHS Signal Word: Warning
EINECSNumber 220-117-2

RUO – Research Use Only