missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine Dihydrate 98.0+%, TCI America™

Catalog No. p-7126321
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06125G 5 g
D061225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06125G Supplier TCI America Supplier No. D06125G
Only null left

Chemical Identifiers

CAS 18835-59-1
Molecular Formula C9H13I2NO5
Molecular Weight (g/mol) 469.01
MDL Number MFCD00150275
InChI Key YWAGQOOMOOUEGY-QVGJJETENA-N
Synonym 3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
PubChem CID 2724330
IUPAC Name (2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate
SMILES O.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O

Specifications

Melting Point 213°C
Quantity 5 g
Formula Weight 432.98
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodo-L-tyrosine Dihydrate