missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diiodo-L-tyrosine Dihydrate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 213°C |
| Quantity | 5 g |
| Formula Weight | 432.98 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,5-Diiodo-L-tyrosine Dihydrate |
Safety and Handling
| TSCA | Yes |