missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-tyrosine dihydrate, 99%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 225610250

Catalog No. AC225610250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

 

Specifications

Specifications

3,5-Diiodo-L-tyrosine dihydrate
Authentic
HOC6H2I2CH2CH(NH2)COOH·2H2O
14,IV,237
-20.7
YWAGQOOMOOUEGY-KLXURFKVSA-N
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate
2724330
99%
18835-59-1
C9H9I2NO3·2H2O
25g
−20.70 (20.00°C c=2.5, 0.1N NaOH/H2O)
3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
C1=C(C=C(C(=C1I)O)I)CC(C(=O)O)N.O.O
469.01
469.01

Chemical Identifiers

18835-59-1
469.01
3,5-diiodo-l-tyrosine dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, 3,5-diiodotyrosine dihydrate, 3,5-dijodtyrosindihydrat, c9h9i2no3.2h2o, 3,5-diiodo-l-tyrosine hydrate, l-tyrosine,3,5-diiodo-, hydrate 1:2, s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoicaciddihydrate, 2s-2-amino-3-4-hydroxy-3,5-diiodophenyl propanoic acid dihydrate, s-2-amino-3-4-hydroxy-3,5-diiodophenyl-propanoic acid dihydrate
(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid;dihydrate
C9H9I2NO3·2H2O
YWAGQOOMOOUEGY-KLXURFKVSA-N
2724330
C1=C(C=C(C(=C1I)O)I)CC(C(=O)O)N.O.O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only