missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-L-thyronine 96.0+%, TCI America™

Catalog No. D1119100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1119100MG 100 mg
D11191G 1 g
D11195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1119100MG Supplier TCI America Supplier No. D1119100MG
Only null left

Chemical Identifiers

CAS 1041-01-6
Molecular Formula C15H13I2NO4
Molecular Weight (g/mol) 525.081
MDL Number MFCD00064987
InChI Key ZHSOTLOTTDYIIK-ZDUSSCGKSA-N
PubChem CID 92859
IUPAC Name (2S)-2-amino-3-[4-(4-hydroxyphenoxy)-3,5-diiodophenyl]propanoic acid
SMILES C1=CC(=CC=C1O)OC2=C(C=C(C=C2I)CC(C(=O)O)N)I

Specifications

Melting Point 256°C
Quantity 100 mg
Formula Weight 525.08
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diiodo-L-thyronine