Learn More
3,5-Dihydroxybenzaldehyde, 98%
CAS: 26153-38-8 | C7H6O3 | 138.12 g/mol
$149.24 - $149.24
Chemical Identifiers
| CAS | 26153-38-8 |
|---|---|
| Molecular Formula | C7H6O3 |
| Molecular Weight (g/mol) | 138.12 |
| MDL Number | MFCD00016611 |
| InChI Key | HAQLHRYUDBKTJG-UHFFFAOYSA-N |
| Synonym | benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde |
| PubChem CID | 94365 |
| ChEBI | CHEBI:50204 |
| IUPAC Name | 3,5-dihydroxybenzaldehyde |
| SMILES | OC1=CC(C=O)=CC(O)=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC425650010
|
Thermo Scientific Chemicals
425650010 |
1 g | Glass bottle |
Each for $149.24
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 26153-38-8 | |
| 138.12 | |
| HAQLHRYUDBKTJG-UHFFFAOYSA-N | |
| 94365 | |
| 3,5-dihydroxybenzaldehyde |
| C7H6O3 | |
| MFCD00016611 | |
| benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde | |
| CHEBI:50204 | |
| OC1=CC(C=O)=CC(O)=C1 |
Specifications
| 26153-38-8 | |
| 100.0 | |
| Gray to White | |
| 97.5% min. (HPLC) | |
| C7H6O3 | |
| MFCD00016611 | |
| benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde | |
| OC1=CC(C=O)=CC(O)=C1 | |
| 138.12 | |
| CHEBI:50204 | |
| 98% | |
| 3, 5-Dihydroxybenzaldehyde |
| 97.5 | |
| 153.0°C to 158.0°C | |
| Authentic | |
| Glass bottle | |
| (HO)2C6H3CHO | |
| 1 g | |
| HAQLHRYUDBKTJG-UHFFFAOYSA-N | |
| 3,5-dihydroxybenzaldehyde | |
| 94365 | |
| 138.12 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye prote
GHS Signal Word: Warning
EINECSNumber : 247-479-4
RUO – Research Use Only