missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dihydroxybenzaldehyde, 98%

Catalog No. p-7022518
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533803 1 g
AAA1533806 5 g
AAA1533814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1533803 Supplier Thermo Scientific Chemicals Supplier No. A1533803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26153-38-8 | C7H6O3 | 138.12 g/mol

3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Air sensitive. Store in cool place. Incompatible with strong acids and strong bases.

Chemical Identifiers

CAS 26153-38-8
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00016611
InChI Key HAQLHRYUDBKTJG-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehyde
PubChem CID 94365
ChEBI CHEBI:50204
IUPAC Name 3,5-dihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1

Specifications

Melting Point 158°C to 161°C
Assay Percent Range 98%
Quantity 1 g
Beilstein 1930147
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol and diethyl ether.
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-479-4
TSCA No
storageNote1 Air sensitive

RUO – Research Use Only