missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dihydroxyacetophenone 98.0+%, TCI America™

Catalog No. D198125G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D198125G 25 g
D198110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D198125G Supplier TCI America Supplier No. D198125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 51863-60-6
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002290
InChI Key WQXWIKCZNIGMAP-UHFFFAOYSA-N
Synonym 3',5'-dihydroxyacetophenone, 1-3,5-dihydroxyphenyl ethanone, 3,5-dihydroxyacetophenone, 5-acetylresorcinol, 1-3,5-dihydroxyphenyl ethan-1-one, unii-kp7r433nnr, ethanone, 1-3,5-dihydroxyphenyl, kp7r433nnr, 1-acetyl-3,5-dihydroxybenzene, 1-3,5-dihydroxyphenyl-ethanone
PubChem CID 103993
IUPAC Name 1-(3,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1)O)O

Specifications

Melting Point 147°C
Quantity 25 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Dihydroxyacetophenone
EINECSNumber (3)-1250
TSCA No