missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dihydroxyacetophenone, 97%

Catalog No. AC208630250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208630250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208630250 Supplier Thermo Scientific Chemicals Supplier No. 208630250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 51863-60-6 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51863-60-6
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002290
InChI Key WQXWIKCZNIGMAP-UHFFFAOYSA-N
Synonym 3',5'-dihydroxyacetophenone, 1-3,5-dihydroxyphenyl ethanone, 3,5-dihydroxyacetophenone, 5-acetylresorcinol, 1-3,5-dihydroxyphenyl ethan-1-one, unii-kp7r433nnr, ethanone, 1-3,5-dihydroxyphenyl, kp7r433nnr, 1-acetyl-3,5-dihydroxybenzene, 1-3,5-dihydroxyphenyl-ethanone
PubChem CID 103993
IUPAC Name 1-(3,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 143.0°C to 147.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (HO)2C6H3COCH3
Quantity 25 g
Beilstein 08, II, 301
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,slightly soluble in acetone
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3', 5'-Dihydroxyacetophenone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 257-480-1

RUO – Research Use Only