missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Dihydroxyacetophenone, 96%

Catalog No. AAB2556709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2556709 10 g
AAB2556718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2556709 Supplier Thermo Scientific Chemicals Supplier No. B2556709

CAS: 51863-60-6 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51863-60-6
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002290
InChI Key WQXWIKCZNIGMAP-UHFFFAOYSA-N
Synonym 3',5'-dihydroxyacetophenone, 1-3,5-dihydroxyphenyl ethanone, 3,5-dihydroxyacetophenone, 5-acetylresorcinol, 1-3,5-dihydroxyphenyl ethan-1-one, unii-kp7r433nnr, ethanone, 1-3,5-dihydroxyphenyl, kp7r433nnr, 1-acetyl-3,5-dihydroxybenzene, 1-3,5-dihydroxyphenyl-ethanone
PubChem CID 103993
IUPAC Name 1-(3,5-dihydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1)O)O

Specifications

Melting Point 142°C to 149°C
Quantity 10 g
Beilstein 1936502
Formula Weight 152.15
Percent Purity 96%
Chemical Name or Material 3',5'-Dihydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 257-480-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only