missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%
Description
3',5-Dihydroxy-4',6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water. Soluble in chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water. Soluble in chloroform and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
Specifications
Specifications
| Melting Point | 190°C to 192°C |
| Quantity | 25 mg |
| Beilstein | 1355109 |
| Merck Index | 14,3901 |
| Solubility Information | Insoluble in water. Soluble in chloroform and methanol. |
| Formula Weight | 344.32 |
| Percent Purity | 97% |
| Chemical Name or Material | 3',5-Dihydroxy-4',6,7-trimethoxyflavone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only