missing translation for 'onlineSavingsMsg'
Learn More

3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%

Catalog No. AAL14495MB
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14495MB 25 mg
AAL14495MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL14495MB Supplier Thermo Scientific Chemicals Supplier No. L14495MB
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 855-96-9 | C18H16O7 | 344.319 g/mol

3',5-Dihydroxy-4',6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.

Chemical Identifiers

CAS 855-96-9
Molecular Formula C18H16O7
Molecular Weight (g/mol) 344.319
MDL Number MFCD00016929
InChI Key KLAOKWJLUQKWIF-UHFFFAOYSA-N
Synonym eupatorin, 3',5-dihydroxy-4',6,7-trimethoxyflavone, eupatorine, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-chromen-4-one, unii-3j474av6my, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-1-benzopyran-4-one, 6-methoxyluteolin 4',7-dimethyl ether, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxychromen-4-one, kinome_3214, spectrum_001037
PubChem CID 97214
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(C(=C(C=C3O2)OC)OC)O)O

Specifications

Melting Point 190°C to 192°C
Quantity 25 mg
Beilstein 1355109
Merck Index 14,3901
Solubility Information Insoluble in water. Soluble in chloroform and methanol.
Formula Weight 344.32
Percent Purity 97%
Chemical Name or Material 3',5-Dihydroxy-4',6,7-trimethoxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only