missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Difluoroacetophenone 97.0+%, TCI America™

Catalog No. p-7127726
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34255G 5 g
D342525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34255G Supplier TCI America Supplier No. D34255G

Chemical Identifiers

CAS 123577-99-1
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.13
MDL Number MFCD00042489
InChI Key OXJLDNSPGPBDCP-UHFFFAOYSA-N
Synonym 3',5'-difluoroacetophenone, 3,5-difluoroacetophenone, 1-3,5-difluorophenyl ethanone, 1-3,5-difluorophenyl ethan-1-one, ethanone, 1-3,5-difluorophenyl, 1-acetyl-3,5-difluorobenzene, 1-3,5-difluoro-phenyl-ethanone, pubchem2179, acmc-1cacd, 3,5-di-fluoroacetophenone
PubChem CID 518596
IUPAC Name 1-(3,5-difluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC(F)=CC(F)=C1

Specifications

Melting Point 40°C
Boiling Point 95°C
Quantity 5 g
Formula Weight 156.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,5′-Difluoroacetophenone