Learn More
Chemical Identifiers
| CAS | 737000-76-9 |
|---|---|
| Molecular Formula | C7H7BF2O3 |
| Molecular Weight (g/mol) | 187.81 |
| MDL Number | MFCD03095353 |
| InChI Key | JXQHRWOUVJRCSR-UHFFFAOYSA-N |
| Synonym | 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid |
| PubChem CID | 2734607 |
| IUPAC Name | (3,5-difluoro-2-methoxyphenyl)boronic acid |
| SMILES | B(C1=CC(=CC(=C1OC)F)F)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC365820010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC365820050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 737000-76-9 | |
| 187.81 | |
| JXQHRWOUVJRCSR-UHFFFAOYSA-N | |
| 2734607 | |
| B(C1=CC(=CC(=C1OC)F)F)(O)O |
| C7H7BF2O3 | |
| MFCD03095353 | |
| 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid | |
| (3,5-difluoro-2-methoxyphenyl)boronic acid |
Specifications
| 737000-76-9 | |
| 100.0 | |
| 96% min. (HPLC) | |
| MFCD03095353 | |
| JXQHRWOUVJRCSR-UHFFFAOYSA-N | |
| (3,5-difluoro-2-methoxyphenyl)boronic acid | |
| 2734607 | |
| 97% |
| 96.0 | |
| Authentic | |
| C7H7BF2O3 | |
| 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid | |
| B(C1=CC(=CC(=C1OC)F)F)(O)O | |
| 187.81 | |
| 187.81 | |
| (3, 5-Difluoro-2-methoxyphenyl)boronic acid, 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only