missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-2-methoxybenzeneboronic acid, 97%

Catalog No. AAL19773MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19773MD 250 mg
AAL1977303 1 g
AAL1977306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL19773MD Supplier Thermo Scientific Chemicals Supplier No. L19773MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 737000-76-9 | C7H7BF2O3 | 187.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 737000-76-9
Molecular Formula C7H7BF2O3
Molecular Weight (g/mol) 187.94
MDL Number MFCD03095353
InChI Key JXQHRWOUVJRCSR-UHFFFAOYSA-N
Synonym 3,5-difluoro-2-methoxyphenyl boronic acid, 3,5-difluoro-2-methoxybenzeneboronic acid, 2-methoxy-3,5-difluorophenylboronic acid, acmc-209orz, 2-methoxy-3,5-difluoro-benzeneboronic acid, boronic acid, 3,5-difluoro-2-methoxyphenyl, boronic acid,b-3,5-difluoro-2-methoxyphenyl, 3,5-difluoro-2-methoxyphenyl boronicacid, 3,5-bis fluoranyl-2-methoxy-phenyl boronic acid, b-3,5-difluoro-2-methoxyphenyl-boronic acid
PubChem CID 2734607
IUPAC Name (3,5-difluoro-2-methoxyphenyl)boronic acid
SMILES COC1=C(F)C=C(F)C=C1B(O)O

Specifications

Melting Point 112°C to 115°C
Quantity 250 mg
Formula Weight 187.94
Percent Purity 97%
Chemical Name or Material 3,5-Difluoro-2-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only