missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine 98.0+%, TCI America™

Catalog No. D05585G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05585G 5 g
D05581G 1 g
D055825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D05585G Supplier TCI America Supplier No. D05585G

Chemical Identifiers

CAS 632-93-9
Molecular Formula C14H21NO4
Molecular Weight (g/mol) 267.33
MDL Number MFCD00005950
InChI Key CDVAIHNNWWJFJW-UHFFFAOYSA-N
Synonym ddc porphyrinogen, 3,5-diethoxycarbonyl-1,4-dihydro-2,4,6-collidine, dicarbethoxydihydrocollidine, 3,5-diethoxycarbonyl-1,4-dihydrocollidine, 3,5-dicarbethoxy-1,4-dihydrocollidine, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-2,4,6-trimethyl-, diethyl ester, ddc van, 3,5-bis ethoxycarbonyl-1,4-dihydrocollidine, 1,4-dihydro-3,5-dicarbethoxycollidine, diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate
PubChem CID 12446
ChEBI CHEBI:83605
IUPAC Name 3,5-diethyl 2,4,6-trimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(C)NC(C)=C(C1C)C(=O)OCC

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Formula Weight 267.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Diethoxycarbonyl-1,4-dihydro-2,4,6-collidine
RTECSNumber US7979375
TSCA Yes