missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorosalicylaldehyde 98.0+%, TCI America™
$36.57 - $142.46
Chemical Identifiers
| CAS | 90-60-8 |
|---|---|
| Molecular Formula | C7H4Cl2O2 |
| Molecular Weight (g/mol) | 191.007 |
| MDL Number | MFCD00003320 |
| InChI Key | FABVMBDCVAJXMB-UHFFFAOYSA-N |
| Synonym | 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro |
| PubChem CID | 66660 |
| IUPAC Name | 3,5-dichloro-2-hydroxybenzaldehyde |
| SMILES | C1=C(C=C(C(=C1Cl)O)C=O)Cl |
Chemical Identifiers
| 90-60-8 | |
| 191.007 | |
| FABVMBDCVAJXMB-UHFFFAOYSA-N | |
| 66660 | |
| C1=C(C=C(C(=C1Cl)O)C=O)Cl |
| C7H4Cl2O2 | |
| MFCD00003320 | |
| 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro | |
| 3,5-dichloro-2-hydroxybenzaldehyde |
Specifications
| 90-60-8 | |
| C7H4Cl2O2 | |
| 5 g | |
| FABVMBDCVAJXMB-UHFFFAOYSA-N | |
| 3,5-dichloro-2-hydroxybenzaldehyde | |
| 66660 | |
| ≥98.0% (GC,T) | |
| 3,5-Dichlorosalicylaldehyde |
| 94°C | |
| MFCD00003320 | |
| 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro | |
| C1=C(C=C(C(=C1Cl)O)C=O)Cl | |
| 191.007 | |
| 191.01 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes