Learn More
3,5-Dichlorosalicylaldehyde, 98%
CAS: 90-60-8 | C7H4Cl2O2 | 191.007 g/mol
$66.56 - $600.21
Chemical Identifiers
| CAS | 90-60-8 |
|---|---|
| Molecular Formula | C7H4Cl2O2 |
| Molecular Weight (g/mol) | 191.007 |
| MDL Number | MFCD00003320 |
| InChI Key | FABVMBDCVAJXMB-UHFFFAOYSA-N |
| Synonym | 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro |
| PubChem CID | 66660 |
| IUPAC Name | 3,5-dichloro-2-hydroxybenzaldehyde |
| SMILES | C1=C(C=C(C(=C1Cl)O)C=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2094106
|
Thermo Scientific Chemicals
B2094106 |
5 g |
Each for $66.56
|
|
|||||
|
AAB2094114
|
Thermo Scientific Chemicals
B2094114 |
25 g |
Each for $187.12
|
|
|||||
|
AAB2094122
|
Thermo Scientific Chemicals
B2094122 |
100 g |
Each for $600.21
|
|
|||||
Description
3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dichlorosalicylaldehyde has been used in the preparation of Schiff base ligands and (RS)-3,5-dichloro-2-[[(1-phenylethyl)imino]methyl]phenol.
Solubility
Insoluble in water. Solubility in methanol is almost transparent.
Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
| 90-60-8 | |
| 191.007 | |
| FABVMBDCVAJXMB-UHFFFAOYSA-N | |
| 66660 | |
| C1=C(C=C(C(=C1Cl)O)C=O)Cl |
| C7H4Cl2O2 | |
| MFCD00003320 | |
| 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro | |
| 3,5-dichloro-2-hydroxybenzaldehyde |
Specifications
| 90-60-8 | |
| C7H4Cl2O2 | |
| 5 g | |
| Air Sensitive | |
| Insoluble in water. Solubility in methanol is almost transparent. | |
| C1=C(C=C(C(=C1Cl)O)C=O)Cl | |
| 191.007 | |
| 191.01 | |
| 3,5-Dichlorosalicylaldehyde |
| 94°C to 97°C | |
| MFCD00003320 | |
| 973391 | |
| 3,5-dichlorosalicylaldehyde, benzaldehyde, 3,5-dichloro-2-hydroxy, 2-hydroxy-3,5-dichlorobenzaldehyde, 3,5-dichloro salicylal, salicylaldehyde, 3,5-dichloro, 3,5-dichloro-2-hydroxy-benzaldehyde, pubchem8246, timtec-bb sbb003642, acmc-209r6w, salicylaldehyde,5-dichloro | |
| FABVMBDCVAJXMB-UHFFFAOYSA-N | |
| 3,5-dichloro-2-hydroxybenzaldehyde | |
| 66660 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 202-005-5
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only