missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloropyridazine 98.0+%, TCI America™

Catalog No. D44471G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44471G Supplier TCI America Supplier No. D44471G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1837-55-4
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD10698048
InChI Key JZSAUQMXKHBZEO-UHFFFAOYSA-N
Synonym pyridazine, 3,5-dichloro, 3,5-dichloro-pyridazine, pyridazine,3,5-dichloro, 3,5-dichlor-pyridazin, acmc-209yx3, ksc538k8n
PubChem CID 19959687
IUPAC Name 3,5-dichloropyridazine
SMILES ClC1=CC(Cl)=NN=C1

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 1 g
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichloropyridazine
TSCA No
Recommended Storage Refrigerator