missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenyl Isocyanate 97.0+%, TCI America™

Catalog No. I030210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I030210G Supplier TCI America Supplier No. I030210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00013859
InChI Key XEFUJGURFLOFAN-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
PubChem CID 94460
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)N=C=O

Specifications

Melting Point 28°C
Color White-Yellow
Boiling Point 244°C
Quantity 10 g
UN Number 2250
Formula Weight 188.01
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dichlorophenyl Isocyanate
EINECSNumber (3)-3940
TSCA Yes