missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenyl isocyanate, 97%

Catalog No. AAA1999106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1999106 5 g
AAA1999114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1999106 Supplier Thermo Scientific Chemicals Supplier No. A1999106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34893-92-0 | C7H3Cl2NO | 188.007 g/mol

3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dichlorophenyl isocyanate is used as an intermediate for medicine and in iprodione. It is also useful for proteomic research.

Solubility
Hydrolysis with water.

Notes
Light and moisture sensitive. Incompatible with water, alcohols, strong bases, amines, acids and strong oxidizing agents.

Chemical Identifiers

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00013859
InChI Key XEFUJGURFLOFAN-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
PubChem CID 94460
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)N=C=O

Specifications

Melting Point 28°C to 32°C
Density 1.38
Boiling Point 242°C to 244°C
Flash Point 134°C (273°F)
Quantity 5 g
UN Number UN2250
Beilstein 510282
Sensitivity Moisture sensitive
Solubility Information Hydrolysis with water.
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichlorophenyl isocyanate
Show More Show Less
Hazard Category H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H330-H334-H318-H315-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P285-P302+P352-P304+P340-P305+P351+P338-P311-P312-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber 252-276-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only