missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenyl isocyanate, 96%, Thermo Scientific™

Chemical Identifiers

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00013859
InChI Key XEFUJGURFLOFAN-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
PubChem CID 94460
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC314140010
SDS View Documents
Thermo Scientific Chemicals
314140010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC314140250
SDS View Documents
Thermo Scientific Chemicals
314140250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

34893-92-0
188.01
XEFUJGURFLOFAN-UHFFFAOYSA-N
94460
C1=C(C=C(C=C1Cl)Cl)N=C=O
C7H3Cl2NO
MFCD00013859
3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
1,3-dichloro-5-isocyanatobenzene

Specifications

34893-92-0
100.0
243.0°C
Authentic
Glass bottle
Cl2C6H3NCO
1g
3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
XEFUJGURFLOFAN-UHFFFAOYSA-N
1,3-dichloro-5-isocyanatobenzene
94460
96%
95.0
1.3800g/mL
110°C
96%
C7H3Cl2NO
MFCD00013859
1.38
Solubility in water: decomposes
C1=C(C=C(C=C1Cl)Cl)N=C=O
188.01
188.01
3, 5-Dichlorophenyl isocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Danger

EINECSNumber : 252-276-9

SDS
Documents

Documents

RUO – Research Use Only