missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorophenyl isocyanate, 96%

Catalog No. AC314140250
Change view
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314140250 25 g Glass bottle
AC314140010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314140250 Supplier Thermo Scientific Chemicals Supplier No. 314140250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 34893-92-0 | C7H3Cl2NO | 188.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34893-92-0
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00013859
InChI Key XEFUJGURFLOFAN-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl isocyanate, 3,5-dichlorophenylisocyanate, benzene, 1,3-dichloro-5-isocyanato, 3,5-dichlorophenyl isocyantae, 1,3-dichloro-5-isocyanato-benzene, 3,5-dichlorobenzenisocyanate, 3,5dichlorophenyl isocyanate, acmc-1ai47, ksc490c1l, 3,5-dichloro-phenylisocyanate
PubChem CID 94460
IUPAC Name 1,3-dichloro-5-isocyanatobenzene
SMILES C1=C(C=C(C=C1Cl)Cl)N=C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 29.0°C to 35.0°C
Color White to Yellow
Density 1.3800g/mL
Boiling Point 243.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula Cl2C6H3NCO
Quantity 25 g
Specific Gravity 1.38
Solubility Information Solubility in water: decomposes
Formula Weight 188.01
Percent Purity 96%
Physical Form Low Melting Solid
Chemical Name or Material 3, 5-Dichlorophenyl isocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Toxic if inhaled.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
EINECSNumber 252-276-9

RUO – Research Use Only