missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™
$136.08 - $486.52
Chemical Identifiers
| CAS | 705-21-5 |
|---|---|
| Molecular Formula | C6H3Cl3O2S |
| Molecular Weight (g/mol) | 245.50 |
| MDL Number | MFCD00051698 |
| InChI Key | RJSQINMKOSOUGT-UHFFFAOYSA-N |
| Synonym | 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t |
| PubChem CID | 522388 |
| IUPAC Name | 3,5-dichlorobenzene-1-sulfonyl chloride |
| SMILES | ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
Chemical Identifiers
| 705-21-5 | |
| 245.50 | |
| RJSQINMKOSOUGT-UHFFFAOYSA-N | |
| 522388 | |
| ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
| C6H3Cl3O2S | |
| MFCD00051698 | |
| 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
| 3,5-dichlorobenzene-1-sulfonyl chloride |
Specifications
| 705-21-5 | |
| C6H3Cl3O2S | |
| 5 g | |
| 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
| ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O | |
| 245.50 | |
| 245.50 | |
| Crystalline Lumps |
| White-Yellow | |
| MFCD00051698 | |
| 3261 | |
| RJSQINMKOSOUGT-UHFFFAOYSA-N | |
| 3,5-dichlorobenzene-1-sulfonyl chloride | |
| 522388 | |
| ≥98.0% (GC,T) | |
| 3,5-Dichlorobenzenesulfonyl Chloride |
Safety and Handling
TSCA : No