Learn More
3,5-Dichlorobenzenesulfonyl chloride, 97%
CAS: 705-21-5 | C6H3Cl3O2S | 245.50 g/mol
$50.84 - $506.64
Chemical Identifiers
| CAS | 705-21-5 |
|---|---|
| Molecular Formula | C6H3Cl3O2S |
| Molecular Weight (g/mol) | 245.50 |
| MDL Number | MFCD00051698 |
| InChI Key | RJSQINMKOSOUGT-UHFFFAOYSA-N |
| Synonym | 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t |
| PubChem CID | 522388 |
| IUPAC Name | 3,5-dichlorobenzenesulfonyl chloride |
| SMILES | ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1100303
|
Thermo Scientific Chemicals
A1100303 |
1 g |
Each for $50.84
|
|
|||||
|
AAA1100306
|
Thermo Scientific Chemicals
A1100306 |
5 g |
Each for $148.45
|
|
|||||
|
AAA1100314
|
Thermo Scientific Chemicals
A1100314 |
25 g |
Each for $506.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 705-21-5 | |
| 245.50 | |
| RJSQINMKOSOUGT-UHFFFAOYSA-N | |
| 522388 | |
| ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O |
| C6H3Cl3O2S | |
| MFCD00051698 | |
| 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
| 3,5-dichlorobenzenesulfonyl chloride |
Specifications
| 705-21-5 | |
| 158°C to 160°C (15 mmHg) | |
| C6H3Cl3O2S | |
| 1 g | |
| 1528657 | |
| 3,5-dichlorobenzene-1-sulfonyl chloride, 3,5-dichlorobenzenesulphonyl chloride, 3,5-dichlorobenzenesulfonylchloride, 3,5-dichloro-benzenesulfonyl chloride, 3,5-dichlorophenyl chlorosulfone, 3,5-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 3,5-dichloro, pubchem5120, acmc-209oey, ksc377a3t | |
| ClC1=CC(=CC(Cl)=C1)S(Cl)(=O)=O | |
| 245.50 | |
| 245.51 | |
| 3,5-Dichlorobenzenesulfonyl chloride |
| 30°C to 33°C | |
| >110°C (230°F) | |
| MFCD00051698 | |
| UN3261 | |
| Moisture sensitive | |
| RJSQINMKOSOUGT-UHFFFAOYSA-N | |
| 3,5-dichlorobenzenesulfonyl chloride | |
| 522388 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only