missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichlorobenzaldehyde 97.0+%, TCI America™

Catalog No. D20121G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20121G 1 g
D20125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20121G Supplier TCI America Supplier No. D20121G

Chemical Identifiers

CAS 10203-08-4
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003352
InChI Key CASRSOJWLARCRX-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dichloro, 3,5-dichloro-benzaldehyde, unii-m74yu7se79, rarechem aq a2 0002, pubchem3068, 3,5-dichlorobezaldehyde, 3,5-dichlorobenaldehyde, 3?5-dichlorobenzaldehyde, benzaldehyde,5-dichloro, acmc-1bxq6
PubChem CID 35746
IUPAC Name 3,5-dichlorobenzaldehyde
SMILES C1=C(C=C(C=C1Cl)Cl)C=O

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 215°C
Quantity 1 g
UN Number 1759
Formula Weight 175.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorobenzaldehyde